Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HCD8MAG15KPX17BK

Millipore

MILLIPLEX® Human CD8+ T Cell Magnetic Bead Panel Premixed 17 Plex Space Saver (Bulk) Packaging

Inflammation/Immunology Bead-Based Multiplex Assays using the Luminex technology enables the simultaneous analysis of multiple cytokine and chemokine CD8+ biomarkers in human serum, plasma and cell culture samples.

Synonym(s):

17-Plex CD8+ T Cell Panel, 17-Plex Magnetic Bead Panel, CD8+ T Cell Magnetic Bead Panel

Sign Into View Organizational & Contract Pricing

Select a Size

96 WELLS
$4,810.00

$4,810.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

Select a Size

Change View
96 WELLS
$4,810.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

$4,810.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

standard curve range: 0.4-1,500 pg/mL
(IL-6)

standard curve range: 0.5-2,000 pg/mL
(TNFα)

standard curve range: 1-3,500 pg/mL
(MIP-1α)

standard curve range: 1-5,000 pg/mL
(Granzyme B)

standard curve range: 1-5,000 pg/mL
(IFNγ)

standard curve range: 1-5,000 pg/mL
(IL-5)

standard curve range: 10-50,000 pg/mL
(Perforin)

standard curve range: 10-50,000 pg/mL
(sFasL)

standard curve range: 2-10,000 pg/mL
(IL-4)

standard curve range: 2-10,000 pg/mL
(sCD137)

standard curve range: 2-7,500 pg/mL
(IL-13)

standard curve range: 2-7,500 pg/mL
(IL-2)

standard curve range: 20-100,000 pg/mL
(Granzyme A)

standard curve range: 4-15,000 pg/mL
(GM-CSF)

standard curve range: 400-1,650,000 pg/mL
(sFas)

standard curve range: 5-20,000 pg/mL
(IL-10)

standard curve range: 7-30,000 pg/mL
(MIP-1β)

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

CD8+ T cells (also known as cytotoxic T cells, cytolytic T cells and killer T cells) belong to a sub-group of T cells capable of inducing the death of infected somatic or tumor cells. T cells that bind weakly to Major Histocompatibility Complex (MHC) self-antigens are positively selected into single-positive CD4+ or CD8+ T cells in the thymus. Those CD8+ T cells that survive and mature after activation become cytotoxic cells, expressing T-cell receptors (TCRs) capable of recognizing specific antigenic peptides bound to Class I MHC molecules and the glycoprotein CD8. Affinity between the CD8 protein and the MHC molecule helps to keep the cytotoxic T cell and target closely bound during antigen specific activation. Once activated, CD8+ T cells are generally classified as having a predefined cytotoxic role within the immune system and undergo IL-2 induced clonal expansion, which increases the number of cells specific for the target antigen. The CD8+ T cells then travel throughout the body in search of antigen-positive somatic/tumor cells.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Application

  • Analytes: GM-CSF, sCD137, IFNγ, sFas, sFasL, Granzyme A, Granzyme B, IL-2, IL-4, IL-5, IL-6, IL-10, IL-13, MIP-1α, MIP-1β, TNF-α, Perforin
  • Recommended Sample type: serum, plasma or tissue/cell lysate and culture supernatant
  • Recommended Sample dilution: Neat
  • Assay Run Time: Overnight
  • Research Category: Inflammation & Immunology

Other Notes

Sensitivity: Refer to kit protocol for sensitivities of individual biomarkers.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service