Skip to Content
MilliporeSigma
All Photos(2)

Documents

HAWG647SP

Millipore

S-Pak® Membrane Filter

mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, white gridded + 3 dispensers & 600pads for microbiological analysis

Synonym(s):

S-Pak 0.45µm 47mm white gridded + 3 dispensers & 600pads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

gridded filter
mixed cellulose esters (MCE)
white filter

Agency

ISO 7704

sterility

sterile

feature

easily wetted with water
hydrophilic
individually wrapped

manufacturer/tradename

S-Pak®

filter diam.

47 mm

pore size

0.45 μm pore size

input

sample type soft drink
sample type sport drink
sample type wine
sample type cosmetics
sample type food(s)
sample type cider
sample type beverage(s)
sample type bottled water
sample type beer

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

S-Pak 0.45μm 47mm white gridded + 3 dispensers & 600pads are made from mixed esters of cellulose and have been optimized for MF method microbiological analysis of water or other liquids. These sterile, ready to use, individually packed filters are easy to use with no toxicity and hydrophobic spots. These filters are been used in the bacteriological analysis of water and to recover bacteria, yeasts and molds from various environmental and filterable samples.

Application

The S-Pak system is suitable for testing drinking, bottled and mineral water according to international standards. These filters find application in Clinical, Cosmetics, Environmental, Food and Beverage, Industrial QC and Pharmaceutical industry.
Microbial analysis of beer, bottled water, cider, sport drink and wine by membrane filtration method.

Linkage

Replaces: HAWG 047 S2

Legal Information

S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service