Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HAWG047BW

S-Pak® Membrane Filter

S-Pak®, filter 0.45 μm, filter diam. 47 mm, circles white mixed cellulose esters (MCE) filter (gridded)

Synonym(s):

MCE Membrane, Mixed Cellulose Filter, Single Pack MCE Membrane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
Pricing and availability is not currently available.

material

circles white mixed cellulose esters (MCE) filter (gridded)

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

manufacturer/tradename

S-Pak®

filter diam.

47 mm

pore size

0.45 μm filter

application(s)

food and beverages
water monitoring

shipped in

ambient

General description

The S-Pak® Membrane Filters are made of Mixed Cellulose esters (MCE), with 0.45μm pore size, white gridded, 47mm diameter, individually packed and protected with a blue sheet of paper which prevents the membrane to encounter the outer film. This state-of-art design inhibits migration of any adhesive on the filter membrane. Thus, ensures no hydrophobic spots on the membrane testing area. The MCE membrane have been manufactured in accordance to our company′s high standards ensuring outstanding recovery of microorganism for the Beverage and water applications.
After the Sample filtration the S-Pak® Membrane is incubated together with a selected culture media which will lead to the recovery of the targeted microorganism.

Application

S-Pak® Membrane Filter is used for microbial filtration of test samples like water, wine, beer, soft drinks from food and beverage industry. Additionally, water samples from pharmaceuticals and other applicable industries can also be tested.

Features and Benefits

  • Ready to use.
  • Sterile.
  • Individually Packed.
  • Higher Recoveries.
  • No Hydrophobic Spots.
  • No Toxicity.

Legal Information

S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service