Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GHRA-88HK

Sigma-Aldrich

Human Ghrelin (ACTIVE) RIA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

grade

for protein analysis

Quality Level

description

ED<sub>80</sub> = 27 ± 7pg/mL ED<sub>50</sub>=125 ± 33 pg/mL ED<sub>20</sub>=652 ± 179 pg/mL
analytes available: Ghrelin
10-2000 pg/mL (standard curve range)

feature

accuracy 83–114%

species reactivity

human

sensitivity measuring range

10 pg/mL

technique(s)

radioimmunoassay: suitable

UniProt accession no.

detection method

radioactive

General description

FUNCTION: SwissProt: Q9UBU3 # Obestatin may be the ligand for GPR39. May have an appetite-reducing effect resulting in decreased food intake. May reduce gastric emptying activity and jejunal motility (By similarity).
SIZE: 117 amino acids; 12911 Da
SUBCELLULAR LOCATION: Secreted.
TISSUE SPECIFICITY: Highest level in stomach. All forms are found in serum as well. Other tissues compensate for the loss of ghrelin synthesis in the stomach following gastrectomy.
PTM: O-octanoylation or O-decanoylation is essential for ghrelin activity. The O-decanoylated forms Ghrelin-27-C10 and Ghrelin-28- C10 differ in the length of the carbon backbone of the carboxylic acid bound to Ser-26. A small fraction of ghrelin, ghrelin-28- C10:1, may be modified with a singly unsaturated carboxylic acid. & Amidation of Leu-98 is essential for obestatin activity (By similarity).
SIMILARITY: SwissProt: Q9UBU3 ## Belongs to the motilin family.

Application

Two-day disequilibrium assay at 4°C. Sample volume: ≤ 100 μL Plasma (collected on ice) recommended

Analysis Note

Linearity 100–142.5%

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Flam. Liq. 3 - STOT RE 2 Inhalation - STOT SE 1

Target Organs

Eyes,Central nervous system, Respiratory Tract

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service