Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF341

Sigma-Aldrich

KGF,
Human Recombinant Animal Free

Synonym(s):

FGF7, HBGF-7, FGF-7, KGF

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$434.00

$434.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
$434.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

$434.00


Please contact Customer Service for Availability

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

dry ice

General description

KGF/FGF-7 is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. KGF/FGF-7 is a mitogen factor specific for epithelial cells and keratinocytes. KGF/FGF-7 signals through FGFR 2b. KGF/FGF-7 plays a role in kidney and lung development, angiogenesis, and wound healing.
Recombinant human KGF/FGF-7 is an 18.9 kDa protein consisting of 163 amino acid residues.
Product Source: Protein is expressed in E.coli.
The protein is manufactured without the use of any animal products/reagents and is considered Animal-Free and can be used in protocols where possible transmisson of animal viruses and contamination must be avoided.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

The ED50 as determined by the dose-dependent stimulation of thymidine uptake by KGF-responsive BaF3 cells is ≤ 10 ng/ml
corresponding to a specific activity of ≥ 1 x 10^5 units/mg.

Physical form

Product is presented in 15 mM Sodium Phosphate pH 7.0 + 0.25 M NaCl and is filtered through a 0.2 micron filter before lyophilization.

Storage and Stability

Store at -20°C for up to 4 months from date of receipt Centrifuge the vial prior to opening.
Reconstitute in 1x PBS to a concentration of 0.5-1.0 mg/ml. Do not vortex. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein and store in working aliquots at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service