Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GF172

Sigma-Aldrich

FGF-10 Protein, Human Recombinant, 25µg

The Fibroblast Growth Factors (FGFs) are heparin binding glycoproteins that exert a variety of biological activities toward cells of mesenchymal, neuronal & epithelial origin.

Synonym(s):

FGF10, Fibroblast Growth Factor 10, Keratinocyte growth factor 2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$447.00

$447.00


Please contact Customer Service for Availability


Select a Size

Change View
25 μG
$447.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

$447.00


Please contact Customer Service for Availability

Quality Level

Assay

>95% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

UniProt accession no.

shipped in

dry ice

General description

The Fibroblast Growth Factors (FGFs) are heparin binding glycoproteins that exert a variety of biological activities toward cells of mesenchymal, neuronal, and epithelial origin. FGF10 belongs to the subgroup of FGFs that also includes FGF3, 7, and 22. Proteins of this family play a central role during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-10 is most related to KGF/FGF-7 and is expressed during the development and preferentially in adult lungs. It signals through the FGFR 2b. Recombinant human FGF-10 is a 19.3 kDa protein consisting of 170 amino acid residues.

Quality

Bioactivity assay #1: The ED50 as determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF− receptors is = 0.5 ng/ml, corresponding to a specific activity of = 2 × 106 units/mg.
Bioactivity assay #2: Determined by a cell proliferation assay using 4MBr-5 cells. The expected−ED50−for this effect is 0.5−15.0 ng/ml.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Human pancreatic cancer organoid biobank (PDAC organoids) with various KRAS mutations to aide in 3D cell culture and cancer research applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service