Skip to Content
MilliporeSigma
All Photos(1)

Documents

GF091

Sigma-Aldrich

Soluble RANK Ligand (sRANKL) Protein, Recombinant human

The recombinant human sRANKL protein is produced in E. Coli.

Synonym(s):

sRANKL, Soluble Receptor Activator of NF-Kappa B, TNFSF11, OPGL, ODF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

>98% (SDS-PAGE and HPLC)

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 ng/μg Endotoxin (1EU/μg)

input

sample type mesenchymal stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... TNFSF11(8600)

General description

Product Source: E. coli
RANKL and RANK are members of the TNF superfamily of ligands and receptors that play an important role in the regulation of specific immunity and bone turnover. RANK (receptor) was originally identified as a dendritic-cell-membrane protein, which by interacting with RANKL augments the ability of dendritic cells to stimulate naïve T-cell proliferation in a mixed lymphocyte reaction, to promote the survival of RANK + T cells, and to regulate T-cell-dependent immune response. RANKL, which is expressed in a variety of cells including osteoblasts, fibroblasts, activated T-cells and bone marrow stromal cells, is also capable of interacting with a decoy receptor called OPG. Binding of soluble OPG to sRANKL inhibits osteoclastogenesis by interrupting the signaling between stromal cells and osteoclastic progenitor cells, thereby leading to excess accumulation of bone and cartilage.
Recombinant human sRANKL produced in E. coli. The protein is a 20.0 kDa polypeptide comprising the TNF homologous region of RANKL (176 amino acid residues).SEQUENCE: MEKAMVDGSW LDLAKRSKLE AQPFAHLTIN ATDIPSGSHK VSLSSWYHDR GWAKISNMTF SNGKLIVNQD GFYYLYANIC FRHHETSGDL ATEYLQLMVY VTKTSIKIPS SHTLMKGGST KYWSGNSEFH FYSINVGGFF KLRSGEEISI EVSNPSLLDP DQDATYFGAF KVRDID

COUNTRY OF ORIGIN: USA

Physical form

Sterile filtered, and then lyophilized from 5 mM sodium phosphate, pH 7.6 + 75 mM NaCl. Reconstitute with distilled water to a concentration of 0.1-1 mg/mL as a stock solution. Solution can then be diluted into other aqueous buffers for immediate use.

Storage and Stability

The lyophilized powder is stable for several weeks at room temperature, but best stored at -20°C for up to one year from date of receipt. Reconstituted stock material can be stored at 2°-8°C for one week, but undiluted aliquots at -20°C are recommended for longer use. Avoid repeated freeze/thaw cycles.

Analysis Note

Specific Activity: Determined by its ability to induce osteoclast formation in RAW264.7 cells using a concentration range of 5.0-10.0 ng/mL.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junchi Huang et al.
Clinical & experimental metastasis, 39(5), 801-814 (2022-08-16)
Metastasis to bone is the leading cause of death from prostate cancer. Interaction between tumor cells and bone cells can promote progression and influence tumor phenotype. It is known that prostate cancer cells support osteoclast differentiation, and degradation of bone

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service