Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G10-013

Millipore

NovAseptic® Mixer Rotor HS-T10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
25171705

material

316L stainless steel head (Mixing Head)
EPDM rubber O-ring
sintered silicon carbide (SSiC) bearing (Female Bearing)

Quality Level

parameter

5-135 °C temp. range (41-275 °C)

storage temp.

2-30°C

General description

Mixing Head: Stainless Steel 316L/EN 1.4435 (Rod: ASME SA-479/EN 10272Plate: ASTM A240/EN 10028-7/EN 10088-2)Female Bearing: Silicone Carbide Ceramic SiC (SsiC)O-ring: EPDM
NovAseptic® mixers offer a complete solution for mixing throughout theprocess line and are designed for a wide variety of mixing applications in thepharmaceutical and biotechnology industries.

Packaging

The Mixing Head is packaged in a closed box.

Components

Mixing Head: Stainless Steel 316L/EN 1.4435 Plate: ASTM A240/EN 10028-7/EN 10088-2)Female Bearing: Silicone Carbide Ceramic SiC (SsiC)O-ring: EPDM

Legal Information

NOVASEPTIC is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service