Skip to Content
MilliporeSigma
All Photos(1)

Documents

FX0450

Supelco

Formic acid

90%, Technical

Synonym(s):

Formic acid, Methanoic Acid,Aminic Acid, Aminic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOOH
CAS Number:
Molecular Weight:
46.03
Beilstein:
1209246
MDL number:
UNSPSC Code:
12352106

product name

Formic Acid, 90%, Technical

vapor density

1.6 (vs air)

Quality Level

vapor pressure

44.8 mmHg ( 20 °C)

Assay

≥90%

form

liquid

autoignition temp.

1004 °F

expl. lim.

57 %

color

APHA: ≤20

refractive index

n20/D 1.370 (lit.)

bp

100-101 °C (lit.)
107 °C/1013 hPa

mp

8.2-8.4 °C (lit.)

transition temp

flash point 71 °C

density

1.22 g/mL at 25 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

OC=O

InChI

1S/CH2O2/c2-1-3/h1H,(H,2,3)

InChI key

BDAGIHXWWSANSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay: 90% min
Color (APHA): 20 max
Form: Clear liquid

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

159.8 °F

Flash Point(C)

71 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service