Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB250A4

Sigma-Aldrich

Milli-Mark® Anti-Bcl-2 Antibody, Alexa Fluor488, clone AW604

clone AW604, Milli-Mark®, from mouse

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$366.00

$366.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
$366.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$366.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AW604, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BCL2(596)

Related Categories

General description

Bcl-2 suppresses apoptosis in a variety of cell systems including factor-dependent lymphohematopoietic and neural cells. Bcl-2 also regulates cell death by controlling the mitochondrial membrane permeability and appears to function in a feedback loop system with caspases. It appears to Inhibit caspase activity either by preventing the release of cytochrome c from the mitochondria and/or by binding to the apoptosis-activating factor (APAF-1). Further understanding in how Bcl-2 functions and interacts with other proteins to promote or inhibit apoptosis may lead to new treatments for cancer, neurological disorders and autoimmune diseases.

Specificity

Antibody recognizes Bcl-2.

Immunogen

KLH-conjugated, synthetic peptide corresponding to human Bcl-2

Application

Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
This Milli-Mark Bcl-2 antibody is validated for use in Flow for the detection of the Milli-Mark Bcl-2 protein.

Quality

Evaulated by flow cytometery using Jurkat cells.

Target description

26 kDa Calculated

Physical form

Protein G Purified
Purified mouse monoclonal IgG2a conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Jurkat Cells

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service