Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB209F

Sigma-Aldrich

Milli-Mark® Anti-CD79β-FITC Antibody, clone SN8

clone SN8, Milli-Mark®, from mouse

Synonym(s):

B-cell antigen receptor complex-associated protein beta chain, Ig-beta, B-cell-specific glycoprotein B29, Immunoglobulin-associated B29 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
$433.00

$433.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 TESTS
$433.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

$433.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

SN8, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD79B(974)

Related Categories

General description

CD79beta is restricted to the B-cell lineage, and is found in pro-B cells before Ig heavy-chain gene rearrangement but after CD79alpha expression. The antibody reacts with an epitope on the extracellular domain of CD79 (Yoshida, 1995). Anti-CD79beta, SN8, was included in the Fifth International Workshop and Conference on Human Leucocyte Differentiation Antigens, and studies by a number of laboratories confirmed its reactivity with CD79beta (Engel, 1995; Persin, 1995).

Specificity

Antibody recognizes Human CD79b antigen.

Application

Milli-Mark Anti-CD79β-FITC Antibody, clone SN8 detects level of CD79β & has been published & validated for use in FC.

Quality

Evaluated by flow cytometry using human lymphocytess

Target description

36 kDa

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service