Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

FCMAB175P

Sigma-Aldrich

Milli-Mark Anti-CD7-PE Antibody, clone CBC.37

clone CBC.37, Milli-Mark®, from mouse

Synonym(s):

T-cell antigen CD7, GP40, T-cell leukemia antigen, TP41, T-cell surface antigen Leu-9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

conjugate:
PE
application:
FACS
clone:
CBC.37, monoclonal
species reactivity:
human
citations:
technique(s):
flow cytometry: suitable
Pricing and availability is not currently available. Contact Technical Service

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CBC.37, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

General description

The CD7 antigen is a membrane-bound 40 kDa glycoprotein expressed on thymocytes, mature T cells, a large majority of natural killer cells, pluripotent hematopoietic stem cells, and progenitor cells of lymphoid and myeloid cells (Bowen, 2002 and 1997). CD7 is one of the earliest T-cell-specific antigens to be expressed in lymphocytes, and the only early marker to persist throughout differentiation (Chang, 1994).

Specificity

Antibody recognizes Human CD7 antigen.

Immunogen

CEM T-cell line (ATTC CCL-199), a T-lymphoblastold cell line.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Milli-Mark Anti-CD7-PE Antibody, clone CBC.37 is validated for use in FC for the detection of CD7.

Quality

Evaluated by flow cytometry using human PBMCs

Target description

40 kDa (Bowen, 2002 and 1997)

Physical form

Protein A purified
Purified Mouse monoclonal IgG2b conjugated to PE in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintained refrigerated at 2-8°C from date of receipt. Protected from light in undiluted aliquots for 6 months from date of receipt.

Analysis Note

Control
PBMCs

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service