Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

EZHEADMM1

EZ-Fit® Manifold Filtration Head

for use with Milliflex Oasis® Funnel, pkg of 1 unit

Synonym(s):

EZ-FIT® Manifold Head, For bioburden application, filtration head for manifold

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
Pricing and availability is not currently available.

material

316L stainless steel head

Sterilization Compatibility

autoclavable compatible

packaging

pkg of 1 unit

manufacturer/tradename

EZ-Fit®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® Funnel

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

The EZ-Fit® manifold head for Milliflex Oasis® funnel allows the use of Milliflex Oasis® funnels with the EZ-Fit® vaccum filtration manifold. The EZ-Fit® filtration unit is a disposable filtration device for bioburden testing and have been designed to ensure optimal microbial recovery. The sterilized, disposable filtration devices streamline the workflow, save time and gives highly reliable microbial enumeration results. The EZ-Fit® head enables easy attachment of the filtration unit to the EZ-Fit® vaccum filtration manifold. The EZ-Fit® Manifold design eliminates sources of contamination that can cause false positive results: all internal areas are accessible and can be cleaned efficiently and all components can be autoclaved.

Application

The EZ-Fit® manifold head for Milliflex Oasis® funnel allows the use of Milliflex Oasis® funnels with the EZ-Fit® vaccum filtration manifold and finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical products analysis.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service