Skip to Content
MilliporeSigma
All Photos(2)

Documents

EZHAGG474

Millipore

EZ-Pak® Membrane Filter

mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 47 mm, gridded green filter, suitable for bioburden testing, input: bottled water, beverage(s)

Synonym(s):

Membrane filters, Semi-permeable membranes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

gridded filter
gridded green filter
mixed cellulose esters (MCE)

Quality Level

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

feature

hydrophilic
individually wrapped

manufacturer/tradename

EZ-Pak®

filter diam.

47 mm

matrix

MF-Millipore

pore size

0.45 μm pore size

input

sample type beer
sample type wine
sample(s) (environmental)
sample type cosmetics
sample type soft drink
sample type cider
sample type food(s)
sample type sport drink
sample type pharmaceutical(s)

sample type bottled water
sample type beverage(s)

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical

compatibility

for use with EZ-Fit® Filtration Unit

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

EZ-Pak Filters are made of Mixed cellulose esters (MCE) with 0.45 μm pore size, green, gridded and 47mm diameter. These filters possess high recoveries and are more appropriate for general microbiological testings. EZ-Pak membranes and dispenser revolutionize filter handling for Membrane Filtration (MF) method testing by combining the benefits of ready-to-use, sterile, individually-packed membranes with unique handling ease.

Application

These filters find application in Cosmetics, Environmental, Food and Beverage, and Industrial QC. Microbial analysis of beer, bottled water, cider, sport drink and wine by membrane filtration method. These filters are widely used to recover yeast and mold from samples

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
EZ-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

A touch-free membrane filter dispenser can streamline your membrane filtration testing workflow. Use with EZ-Pak® filter membranes for high-volume sampling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service