Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EZFITEFUN3

Millipore

EZ-Fit® Manifold

3-place, For EZ-Fit® Filtration units

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

material

stainless steel

Sterilization Compatibility

autoclavable compatible

manufacturer/tradename

EZ-Fit®

input

sample type cosmetics
sample type beer
sample type bottled water
sample type pharmaceutical(s)

application(s)

bioburden testing
cosmetics
environmental
food and beverages
water monitoring

shipped in

ambient

General description

EZ-Fit® Manifold′s unique design optimizes workflow and maintenance and reduces the risk of contamination. Different filtration heads fit both reusable and disposable filtration devices. Low height design increases comfort when used in laminar flow hoods. EZ-Fit® Manifold prevents biofilm formation because every single component can be removed for easy cleaning and autoclaving.

Application

The EZ-Fit® filtration Manifold is used for universal laboratory filtrations and supports the simultaneous filtration of three test samples. The manifold is suitable to be used with EZ-Fit® filtration units.

Preparation Note

Sterilization Method
Autoclavable

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service