Key Documents
EZFDRAW01
Millipore
EZ-Fluo Drawer
For EZ-Fluo Reader rapid microbial detection
About This Item
UNSPSC Code:
41116500
NACRES:
NB.79
Pricing and availability is not currently available.
material
aluminum
description
removable
manufacturer/tradename
EZ-Fluo
application(s)
bioburden testing
food and beverages
microbiology
pharmaceutical
compatibility
for use with EZ-Fluo system
General description
EZ-Fluo Drawer is designed to hold the Petri-Pad containing the test sample during the reading.
The drawer can be removed easily and replaced without any specific tools.
The EZ-Fluo system can detect and quantify microbial contamination in any filterable sample. The method used is fast, non-destructive, and fluorescent staining based. The non-destructive feature allows the user to re-incubate the filtered sample for growth on non-selective microbial media.
Application
EZ-Fluo Drawer is used to hold test samples from food and pharmaceutical industries to check for live microbial activity.
Legal Information
EZ-FLUO is a trademark of Merck KGaA, Darmstadt, Germany