Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EX0566

Supelco

Ethylene glycol

Synonym(s):

2-Ethanediol,1,2-ethanediol

Sign Into View Organizational & Contract Pricing


About This Item

vapor pressure

0.053 hPa ( 20 °C)

Quality Level

impurities

≤0.0001% Chloride (Cl)
≤0.005% Acidity (as acetic acid)
≤0.03% Water (H2O)

ign. residue

≤0.005%

pH

6-7.5 (20 °C, 100 g/L in H2O)

bp

197.6 °C/1013 hPa

mp

-14--10 °C

transition temp

flash point 111 °C

solubility

1000 g/L

storage temp.

room temp

Application

anhydrous, Reagent

Analysis Note

Acidity (as acetic acid):0.005% max
Boiling range, begin:195°C min
Boiling range, end:198°C max
Chloride (Cl):0.0001% max
Residue on ignition:0.005% max
Solubility in water:Passes test
Specific Gravity at 20/20°C:1.114-1.117
Water (H2O):0.03% max

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service