Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EX0564

Supelco

Ethylene glycol

Synonym(s):

Ethylene Glycol, 1,2-Ethanediol, 1

Sign Into View Organizational & Contract Pricing

Select a Size

3 L
$614.00

$614.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
3 L
$614.00

About This Item

Empirical Formula (Hill Notation):
C2H6O2
CAS Number:
Molecular Weight:
62.07
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-473-3
bp:
197.6 °C/1013 hPa
vapor pressure:
0.053 hPa ( 20 °C)

$614.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.053 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

impurities

≤0.2% Water (H2O)

color

APHA: ≤10

pH

6-7.5 (20 °C, 100 g/L in H2O)

bp

197.6 °C/1013 hPa

mp

-14--10 °C

transition temp

flash point 111 °C

solubility

1000 g/L

density

1.11 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

OCCO

InChI

1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2

InChI key

LYCAIKOWRPUZTN-UHFFFAOYSA-N

Analysis Note

Assay (GC): 99.0% min
Color (APHA): 10 max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Water (H2O): 0.2% max

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Oral

Target Organs

Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - open cup

Flash Point(C)

115 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service