Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EX0275

Supelco

Specially Denatured Alcohol 3C

200 Proof

Synonym(s):

ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352108

Quality Level

form

liquid

concentration

200 proof in alcohol (denatured)

transition temp

flash point 13 °C

shipped in

ambient

storage temp.

room temp

Application

200 Proof

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service