Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ECM534

Sigma-Aldrich

β Integrin-mediated Cell Adhesion Array Kit, fluorimetric

The Beta Integrin-Mediated Cell Adhesion Array Kit can be used for assessing the presence or absence of specific integrins on the cell surface.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$850.00

$850.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
$850.00

About This Item

UNSPSC Code:
12352207
eCl@ss:
32161000
NACRES:
NA.32

$850.00


Please contact Customer Service for Availability

Quality Level

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

cell based assay: suitable

detection method

fluorometric

shipped in

wet ice

Gene Information

human ... ITGB1(3688)

Application

Research Category
Cell Structure
The Beta Integrin-Mediated Cell Adhesion Array Kit can be used for assessing the presence or absence of specific integrins on the cell surface.
The CHEMICON® Beta Integrin-Mediated Cell Adhesion Array Kit can be used for assessing the presence or absence of specific integrins on the cell surface. This assay can substitute for FACS analysis (6) and is useful for screening human adherent cells for cell surface integrins or for monitoring in vitro cell differentiation or genetic modification of cells.



For Research Use Only. Not for use in diagnostic procedures.

Packaging

96 wells

Components

Beta Integrin Array Plate: (PN: 90600) One 96-well plate with 12 strips. Each strip contains one well each of the mouse anti-beta integrin monoclonal antibodies: (beta1, beta2, beta3, beta4, beta6, alphaVbeta5, and alpha5beta1), and one goat anti-mouse negative well. See the plate layout on insert data sheet.

4X Cell Lysis Buffer: (Part No. 90130) One bottle - 16 mL

CyQuant GR Dye: (Part No. 90132) One vial - 75 μL

Assay Buffer: (Part No. 90601) One bottle - 100 mL

Storage and Stability

The Beta Integrin Array Plate can be stored at 2° to 8°C in the foil pouch up to its expiration date. Unused strips may be placed back in the pouch for storage. Ensure that the desiccant remains in the pouch, and that the pouch is securely closed. Keep the remaining kit components at 2° to 8°C.

Precautions

No data is available on the biological toxicity of CyQuant GR dye. This reagent binds to nucleic acids, and as such, should be treated as a potential mutagen which may cause cancer and heritable genetic damage. Handle with caution. The DMSO stock solution should be handled with special caution as DMSO can facilitate the entry of organic molecules into tissues.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
GR is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Rath et al.
Cellular and molecular life sciences : CMLS, 79(6), 340-340 (2022-06-07)
Cerebral cavernous malformations (CCM) are low-flow vascular lesions prone to cause severe hemorrhage-associated neurological complications. Pathogenic germline variants in CCM1, CCM2, or CCM3 can be identified in nearly 100% of CCM patients with a positive family history. In line with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service