Skip to Content
MilliporeSigma
All Photos(1)

Documents

DX2519

Supelco

Drierite Indicating

size 10-20 mesh

Synonym(s):

desiccant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123003

form

granular

size

10-20 mesh

color

blue

particle size

10-20 mesh

shipped in

ambient

storage temp.

room temp

General description

10-20 Mesh

Analysis Note

Description: Blue irregular granules

Legal Information

Drierite is a trademark of W.A. Hammond Drierite Co., LTD

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service