Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DX2091

Supelco

p-Dioxane

OmniSolv®

Synonym(s):

p-Dioxane, 1,4-Dioxane, 1

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$507.00
4 L
$1,020.00

$507.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
$507.00
4 L
$1,020.00

About This Item

Linear Formula:
-O(CH2)2O(CH2)2-
CAS Number:
MDL number:
UNSPSC Code:
12191504
EC Index Number:
204-661-8
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
101.5 °C/1013 hPa
vapor pressure:
41 hPa ( 20 °C)

$507.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

41 hPa ( 20 °C)

Quality Level

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

shelf life

12 mo. (from manufacture date)

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.05% Water (H2O)
≤2 mg/L Peroxide (as H2O2)

evapn. residue

≤1 mg/L

color

clear colorless,





refractive index

n20/D 1.4206-1.4226

pH

6-8 (20 °C, 500 g/L in H2O)

bp

101.5 °C/1013 hPa

mp

12 °C

transition temp

flash point 11 °C

density

1.03 g/cm3 at 20 °C

UV cutoff

≤215 nm

UV absorption

λ: 215 nm Amax: ≤1.00
λ: 250 nm Amax: ≤0.300
λ: 300 nm Amax: ≤0.020
λ: 350 nm Amax: ≤0.005
λ: 400 nm Amax: ≤0.005

shipped in

ambient

storage temp.

no temp limit

SMILES string

O1CCOCC1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Application

OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Expiration dateExp 12 months from mfg dateAssay (GC)99.9% minBoiling range101-102°CEvaporation residue1mg/L maxFiltered through 0.2 µm filterTo pass testPeroxide (as H2O2)2mg/L maxRefractive index (n 20/D)1.4206-1.4226UV Abs. at 215 nm1.00AU maxUV Abs. at 250 nm0.300AU maxUV Abs. at 300 nm0.020AU maxUV Abs. at 350 nm0.005AU maxUV Abs. at 400 nm0.005AU maxUV Cut-off215nm maxWater (H2O)0.05% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service