Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DR1106

Sigma-Aldrich

Anti-MSH6 Mouse mAb (1F2)

liquid, clone 1F2, Calbiochem®

Synonym(s):

Anti-MutS Homolog 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F2, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MSH6(2956)

General description

Purified mouse monoclonal antibody. Recognizes the ~140-150 kDa MSH6 protein.
Recognizes the ~140-150 kDa MSH6 protein in HeLa cells and human kidney tissue.
This Anti-MSH6 Mouse mAb (1F2) is validated for use in ELISA, Immunoblotting, Immunocytochemistry for the detection of MSH6.

Immunogen

A recombinant polypeptide corrresponding to amino acids of 931-1031 human MSH6, expressed as a GST fusion protein

Warning

Toxicity: Regulatory Review (Z)

Analysis Note

Positive Control
Human kidney tissue, HeLa cells

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service