Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DR1093

Sigma-Aldrich

Anti-ETV6 Mouse mAb (3B10)

liquid, clone 3B10, Calbiochem®

Synonym(s):

Anti-ETS Variant 6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$594.00

$594.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$594.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$594.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B10, monoclonal

form

liquid

does not contain

preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ETV6(2120)

General description

Purified mouse monoclonal antibody. Recognizes the ~55-60 kDa ETV6 protein.
Recognizes the ~55-60 kDa ETV6 protein in Jurkat cells and human placenta tissue.
This Anti-ETV6 Mouse mAb (3B10) is validated for use in ELISA, Immunoblotting, Paraffin Sections for the detection of ETV6.

Immunogen

Full-length, recombinant, human ETV6 (aa 1-453), expressed as a GST fusion protein

Application



ELISA (see comments)
Immunoblotting (1-5 g/ml)
Paraffin Sections (3 g/ml)

Warning

Toxicity: Regulatory Review (Z)

Physical form

In PBS, pH 7.2.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Jurkat cells, Human placenta tissue

Other Notes

For ELISA, this antibody can be used as the capture antibody; recommended concentration is 10 ng/ml, 1 µg/well, 100 µl total volume. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service