Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

DF-041

Sigma-Aldrich

DMEM/F12, with HEPES, L-Glutamine

The DMEM/F12, with HEPES, L-Glutamine is available in a 500 mL format & has been optimized & validated for Stem cell culture.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

cell culture | mammalian: suitable
cell culture | stem cell: suitable

input

sample type epithelial cells
sample type: mouse embryonic stem cell(s)
sample type: human embryonic stem cell(s)
sample type cardiac stem cell(s)
sample type pancreatic stem cell(s)
sample type hematopoietic stem cell(s)
sample type mesenchymal stem cell(s)
sample type induced pluripotent stem cell(s)
sample type neural stem cell(s)

components

NaHCO3: 1200.00 mg/L
phenol red: 8.10 mg/L
L-glutamine: 365.00 mg/L
glucose: 3151.00 mg/L (Dextro)
HEPES: 3574.50 mg/L

Physical form

Contains L-Glutamine and HEPES.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Skip weekend feedings. Defined serum-free and feeder-free expansion media for human pluripotent stem cells (ES and iPS cells). See publications and protocols.

The Simplicon™ RNA Reprogramming Technology is a next generation reprogramming system that uses a single synthetic, polycistronic self-replicating RNA strand engineered to mimic cellular RNA to generate human iPS cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service