Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CX2335

Supelco

Cyclohexanone

Meets ACS Specifications GR ACS

Synonym(s):

Cyclohexanone, Pimelic Ketone

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$262.00
4 L
$758.00
20 L
$2,230.00
200 L
$11,580.00

$262.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 ML
$262.00
4 L
$758.00
20 L
$2,230.00
200 L
$11,580.00

About This Item

Linear Formula:
-CH2(CH2)4C(=O)-
CAS Number:
MDL number:
UNSPSC Code:
12191502
EC Index Number:
203-631-1
grade:
ACS reagent
bp:
155 °C/1013 hPa
vapor pressure:
4.5 hPa ( 20 °C)

$262.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

ACS reagent

Quality Level

vapor pressure

4.5 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

shelf life

36 mo. (from receipt date)

dilution

(for analytical testing)

impurities

≤0.05% Water (H2O)

evapn. residue

≤0.05%

color

APHA: ≤10, at time of manufacture

pH

7 (20 °C, 70 g/L in H2O)

bp

155 °C/1013 hPa

mp

-31 °C

transition temp

flash point 43 °C

solubility

90 g/L

density

0.95 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

O=C1CCCCC1

InChI

1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2

InChI key

JHIVVAPYMSGYDF-UHFFFAOYSA-N

Application

Meets ACS Specifications GR ACS

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Expiration date: Exp 36 months from receipt date
Assay (GC): 99.0% min
Color (APHA) at time of manufacture: 10 max
Evaporation residue: 0.05% max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Water (H2O): 0.05% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

111.2 °F - closed cup

Flash Point(C)

44 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service