Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CX0855

Supelco

Chlorobenzene

Meets ACS Specifications GR ACS

Synonym(s):

Chlorobenzene, Monochlorobenzene,Phenylchloride, Benzene Chloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 LB
$6,950.00

$6,950.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 LB
$6,950.00

About This Item

Empirical Formula (Hill Notation):
C6H5Cl
CAS Number:
Molecular Weight:
112.56
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-628-5
Assay:
≥99.5% (GC)
bp:
132 °C/1013 hPa
vapor pressure:
12 hPa ( 20 °C)

$6,950.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

12 hPa ( 20 °C)

Quality Level

Assay

≥99.5% (GC)

form

liquid

impurities

≤0.004 meq/g Titrable acid

evapn. residue

≤20 ppm

color

APHA: ≤30

pH

7 (20 °C)

bp

132 °C/1013 hPa

mp

-45 °C

transition temp

flash point 27 °C

solubility

0.207 g/L

density

1.11 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

Clc1ccccc1

InChI

1S/C6H5Cl/c7-6-4-2-1-3-5-6/h1-5H

InChI key

MVPPADPHJFYWMZ-UHFFFAOYSA-N

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay (GC): 99.5% min
Color (APHA): 30 max
Evaporation residue: 20ppm max
Form: Clear liquid
Titrable acid: 0.004meq/g max

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service