Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CS204279

Sigma-Aldrich

EF1α-STEMCCA (OKSM) Lentivirus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service