Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CH2900019

SAFC

Metafolin® ( L-5-methyltetrahydrofolic acid calcium (L-5-MTHF-Ca))

L-5-methyltetrahydrofolic acid calcium (L-5-MTHF-Ca), USP

Pharma Manufacturing

Synonym(s):

(6S)-5-Methyl-5,6,7,8-tetrahydropteroyl-L-glutamic acid, (6S)-5-Methyltetrahydrofolic acid, Calcium salt, Calcium salt [(6S)-5-MTHF-Ca], L-5-MTHF; L-5-MTHF-Ca, L-5-methyltetrahydrofolate, L-5-methyltetrahydrofolate-Calcium, L-5-methyltetrahydrofolic acid (L-5-MTHF-Ca)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

GMO free
GMP
Halal
Kosher

Agency

USP

form

powder

application(s)

pharmaceutical
solid formulation
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Metafolin® can be used as dietary supplement for human use with a daily dosage of 200 – 1000 µg.

Packaging

Customized packaging possible

Legal Information

METAFOLIN is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Arcofolin® Methylfolate as a superior alternative to Folic Acid has uniquely become available for nutritional supplementation and fortification.

Medicine for children poses unique formulation challenges compared to adults. Consider developmental physiology and age specifics when designing pharmaceuticals. Quality issues can severely impact patient safety. Therefore, excipient quality, supplier selection, and supply chain security are crucial, particularly for pediatric formulations.

Related Content

Our high-quality folates are manufactured under cGMP guideline ICH Q7 and backed by dedicated regulatory support worldwide.

Developing formulations specifically for infants and children is increasingly important. To help you master these challenges effectively, we offer an extensive portfolio of well-established high-quality excipients and APIs for pediatric pharmaceutical formulations that are proven in practice.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service