Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

CFSP001700

Millipore

Cellulose Absorbent Sample Pads

thickness 0.83 mm, filter W × L 17 mm × 100 m, pack of 1 roll

Synonym(s):

Absorbent pad, C083 Cellulose Fiber Sample Pad Roll, Cellulose pad, Sample pad

Sign Into View Organizational & Contract Pricing

Select a Size

1 ROLL
$81.80

$81.80


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
1 ROLL
$81.80

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

$81.80


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

packaging

pack of 1 roll

Quality Level

manufacturer/tradename

SureWick®

L

100 m (328 ft)

W

17 mm

filter W × L

17 mm × 100 m

thickness

0.83 mm

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

SureWick® Pad Materials are designed and optimised for use in rapid diagnostic lateral flow tests that require superior consistency and performance. SureWick® cellulose fiber pad materials are used as sample and absorbant pads in lateral flow assays. This pack contains one 17mm x 100m roll of C083 cellulose fiber pad material.

Application

SureWick® Pad Materials are optimised for use as sample, conjugate, or absorbent pads in a broad range of Lateral Flow test applications, including Infectious Disease testing (COVID-19, HIV, Influenza, Malaria, STDs etc.), as well as in women’s health, biomarker detection, drugs of abuse testing, drug development, environmental testing, agricultural applications, food safety, product quality testing, and animal health.

Features and Benefits

  • SureWick® cellulose fiber pads are made from non-woven, 100% pure cellulose fiber.
  • These pads do not contain binders or glues, which can interfere with assay performance.
  • Manufactured to ensure excellent consistency, they provide reliable, predictable sample flow.
  • They exhibit consistent thickness, bed volume, and uniform surface quality.
  • Quick delivery and lead time.
  • Backed by excellent technical support.

Legal Information

SUREWICK is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shima Dalirirad et al.
Analytical biochemistry, 596, 113637-113637 (2020-02-23)
A lateral flow assay using DNA aptamer-based sensing for the detection of dopamine in urine is reported. Dopamine duplex aptamers (hybridized sensor with capture probe) are conjugated to 40-nm Au nanoparticles (AuNPs) with 20T linkers. The detection method is based

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service