Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CC105

Sigma-Aldrich

Human Integrin α5β1 Protein, Triton X-100 Formulation

Synonym(s):

VLA-5

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$534.00
50 μG
$792.00

$534.00


Please contact Customer Service for Availability


Select a Size

Change View
25 μG
$534.00
50 μG
$792.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

$534.00


Please contact Customer Service for Availability

biological source

human

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

impurities

CONTAMINANT(S)
none detected

NCBI accession no.

UniProt accession no.

Gene Information

human ... ITGA5(3678)

General description

Integrin a5/b1 purified from smooth muscle and placental tissue under nondenaturing conditions by affinity chromatography on fibronectin-Sepharose (Belkin, 1990).



Two main protein bands corresponding to a5 (145 kD) and beta1 (120 kD) subunits are seen in silver stained 7.5%

polyacrylamide gels under nonreduced conditions. Minor protein band at the 210 kD corresponding to beta1 subunit disulfide dimmer, which is formed during electrophoresis and disappears after reduction.
Product Source: Human smooth muscle and placental tissue.

Application

Electrophoresis and Immunoblotting control. Due to the existance of reduction sensitive sites within the integrin subunits, it is recommended that gels be run under non-reducing conditions for best results. Under non-reducing conditions, the alpha5 subunit will migrate at 155 kDa while the beta1 subunit will migrate at 110 kDa.

Function: Liposomes containing Integrin alpha5/beta1 adhere to surfaces coated with fibronectin in an RGD-dependent fashion.

Physical form

Purified protein in 20 mM Tris-HCl, pH 7.5, 150 mM NaCl, 2 mM MgCl2 containing 0.2% Triton X-100. No preservatives. Source material for the enclosed purified human proteins, Integrin alpha5beta1, has been tested for antibodies to HIV, HCV, and HbsAg and found to be negative by an approved test.

Storage and Stability

Maintain at -70°C in undiluted aliquots. Avoid repeated freeze/thaw cycles.

Analysis Note

Nonreduced SDS-PAGE of 15 mg on 5-15% polyacrylamide slab gel and staining with Coomassie shows only two main bands at 110 kDa (beta1 subunit) and 155 kDa (alpha5 subunit).A weak band at 205 kDa is due to dimerization of the b1 subunit (Belkin et al., 1996)

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service