Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BX1632

Supelco

Buffer pH 7.0 Yellow

Synonym(s):

buffer solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

analytical standard

Quality Level

Agency

traceable to NIST

pH

7 ( in H2O)

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

shipped in

ambient

storage temp.

15-25°C

General description

Buffer solution pH 7.0 yellow is an off-the-shelf buffer solution used in standardized method applications such as pH calibration.

Application

This ready-to-use pH 7.0 yellow buffer solution comprising the phosphate mixture is suitable for maintaining the pH of the solution or use in pH meter calibration.[1]

Features and Benefits

  • Custom-tailored solution for a cost-effective process with minimized risk
  • Stored in a plastic bottle equipped with a convenient squirt top dispensing cap
  • Traceable to NIST standards
  • Safety labeled
  • Recommended for use in general chemistry applications

Specifications

Tolerance: ±0.01

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of protein and solution properties on the Donnan effect during the ultrafiltration of proteins
Bolton GR, et al.
Biotechnology Progress (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service