Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BX1160

Supelco

Bromocreasol green sodium salt

pH indicator, meets analytical specifications of USP/NF GR, ACS

Synonym(s):

Bromocresol Green, Sodium Salt, 3`,3``,5`,5``-tetrabromo-m-cresolsulfonephthalein Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H13Br4NaO5S
CAS Number:
Molecular Weight:
720.00
MDL number:
UNSPSC Code:
12352107
EC Index Number:
263-657-4

Product Name

Bromocresol Green, Sodium Salt, meets analytical specifications of USP/NF GR ACS

Agency

meets analytical specifications of USP/NF GR ACS

form

powder

color

brown to green, variable

visual transition interval

3.8-5.4, yellow to blue

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C21H14Br4O5S.Na/c1-9-12(7-14(22)19(26)17(9)24)21(13-8-15(23)20(27)18(25)10(13)2)11-5-3-4-6-16(11)31(28,29)30-21;/h3-8,26-27H,1-2H3;/q;+1/p-1

InChI key

HEFSGAHJDGZCHA-UHFFFAOYSA-M

General description

Meets ACS Specifications GR ACS

Application

Meets ACS Specifications GR ACS

Analysis Note

Clarity of solutionTo pass testColorVariable (brown to green)FormPowderVisible spectrum, max. absorbance0.80AU minVisible spectrum, max. wavelength615-620nmVisual transition (from pH 3.8 to pH 5.4)Yellow to Blue

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service