Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

B311-200DS

Tryptic Soy Broth Ready-to-use Bottles

bottle capacity 300 mL, bottle filling volume 200 mL, closure type, blue crimp cap with septum, box of 12 bottles

Synonym(s):

CASO Broth, Casein Soy Broth, Casein Soyabean Digest Broth, TSB, Tryptone Soy Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

material

round glass bottle

Quality Level

Agency

EP 2.6.13 (
)

JP 4.06
USP 71

sterility

sterile; autoclaved

form

liquid

feature

closure type blue crimp cap with septum

packaging

box of 12 bottles

technique(s)

microbiological culture: suitable
sterility testing: suitable

bottle capacity

300 mL

bottle filling volume

200 mL

pH

7.3±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tryptic Soy Broth is a nutrient-rich universal medium free of inhibitors and targets to enumerate a broad spectrum of bacteria, yeast, and molds. The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source, which promotes the growth of the microflora. The broth medium is used during sterility testing due to its wide range of specificity.

Application

Tryptic Soy broth is used to cultivate and enumerate a broad range of bacteria, yeast and fungi present in a test sample.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service