Skip to Content
MilliporeSigma
All Photos(1)

Documents

AX1699KC

Supelco

Solvent KC

AQUASTAR®

Synonym(s):

Karl Fischer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601

Quality Level

form

liquid

transition temp

flash point 55.6 °C

shipped in

ambient

storage temp.

room temp

Application

AQUASTAR
Contains dichloromethane for enhanced solubility of oils and fats when testing aldehydes and ketones.
AX1699KC-6 is packaged in 38/430 glass bottles
AX1699KC-5 is packaged in 38/430 poly coated glass bottles

Analysis Note

Color: Colorless
Form: Clear liquid
Performance: To pass test
Water (H2O): 0.06-0.10%

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Immune system, thymus

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

132.1 °F

Flash Point(C)

55.6 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service