Skip to Content
MilliporeSigma
All Photos(1)

Documents

AX1697AN

Supelco

Coulomat A

Safer Formulation for Moisture Determination AQUASTAR®

Synonym(s):

Karl Fischer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601

form

liquid

transition temp

flash point 7 °C

shipped in

ambient

storage temp.

room temp

Application

Pyridine-free and Chloroform-free.
AX1697AN-1 and AX1697AN-30 are packaged in 38/430 poly coated glass bottles.
AX1697AN-4 is packaged in 38/430 glass bottles.

Analysis Note

Color: Colorless to light yellow
Form: Clear liquid
Performance: To pass test
Water (H2O): 1-50ppm

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.6 °F

Flash Point(C)

7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service