Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AX1270

Supelco

Ammonium Chloride

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Ammonium Chloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$122.00
12 KG
$667.00

$122.00


Available to ship onApril 03, 2025Details


Request a Bulk Order

Select a Size

Change View
500 G
$122.00
12 KG
$667.00

About This Item

Empirical Formula (Hill Notation):
H4ClN
CAS Number:
Molecular Weight:
53.49
MDL number:
UNSPSC Code:
12352301
EC Index Number:
235-186-4
grade:
ACS reagent
form:
crystals

$122.00


Available to ship onApril 03, 2025Details


Request a Bulk Order

grade

ACS reagent

Quality Level

Agency

meets USP/NF

vapor pressure

1.3 hPa ( 30 °C)

Assay

≥99.5% (silver nitrate titration)

form

crystals

impurities

≤0.005% Insoluble matter

ign. residue

≤0.01%

pH

4.7 (25 °C, 200 g/L in H2O, External MSDS)

mp

338 °C (sublimed; External MSDS)

anion traces

phosphate (PO43-): ≤2 ppm
sulfate (SO42-): ≤0.002%

cation traces

Ca: ≤0.001%
Fe: ≤2 ppm
Mg: ≤5 ppm
heavy metals: ≤5 ppm

shipped in

ambient

storage temp.

room temp

SMILES string

Cl[NH4]

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay (argentometric)99.5% minCalcium (Ca)0.001% maxFormWhite crystalsHeavy Metals (by ICP-OES)5ppm maxIdentificationTo pass testInsoluble matter0.005% maxIron (Fe)2ppm maxMagnesium (Mg)5ppm maxPhosphate (PO4)2ppm maxResidue after ignition0.01% maxSulfate (SO4)0.002% maxpH of a 5% solution at 25°C4.5-5.5

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service