Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ARK2210

SAFC

Ethanol

API Grade

Pharma Manufacturing

Synonym(s):

Absolute Alcohol, Alcohol, EtOH, Ethanol - API Grade, Ethyl Alcohol, Ethyl Hydrate, Ethyl Hydroxide, Ethylic Alcohol, Hydroxyethane, Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
12191601
E Number:
E1510
EC Index Number:
603-002-00-5
Pricing and availability is not currently available.

grade

GMP

Quality Level

Agency

Ph. Eur.
USP

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

form

liquid

autoignition temp.

683 °F

expl. lim.

19 %, 60 °F

impurities

low in endotoxins

refractive index

n20/D 1.3600 (lit.)

bp

78 °C (lit.)

mp

−114 °C (lit.)

application(s)

parenterals
parenterals
pharmaceutical

format

neat

storage temp.

2-25°C

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Searching for an API that’s right for you? Our high-quality APIs provide all the necessary regulatory information for your needs. We offer best-practice support throughout your entire registration process and boast an excellent track record facilitating our customers’ compliance with international standards – making us your preferred global partner for APIs.
Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Ethanol is used in some drug product formulations where the Ethanol has a pharmacological effect and has direct patient contact or contact with other substances with direct patient contact. In these cases, there is a need for Ethanol which is fully compliant with ICH Q7 GMP requirements for APIs.
Our Ethanol API supports our customers with their final drug product compliance. The drug products containing Ethanol API are used for example as antiseptic agents and antidotes, as well as for alcohol septal ablation to treat cardiomyopathy and several oncology applications like “Alcoholneurolysis”, “Ethanol Ablation of Hepatocellular Carcinoma” and “Percutaneous ethanol ablation”.

Packaging

ARK2210-1l / Glass Bottle

ARK2210-5l / Stainless Steel Drum

ARK2210-25l / Stainless Steel Drum

ARK2210-100l / Stainless Steel Drum

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service