Skip to Content
MilliporeSigma
All Photos(1)

Documents

APT750

Sigma-Aldrich

ApopNexin Annexin V FITC Apoptosis Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.45

species reactivity

mammals

Quality Level

technique(s)

flow cytometry: suitable

UniProt accession no.

detection method

fluorometric

General description

FUNCTION: SwissProt: P08758 # This protein is an anticoagulant protein that acts as an indirect inhibitor of the thromboplastin-specific complex, which is involved in the blood coagulation cascade.
SIZE: 320 amino acids; 35937 Da
SUBUNIT: Monomer. Binds ATRX (By similarity).
DOMAIN: SwissProt: P08758 A pair of annexin repeats may form one binding site for calcium and phospholipid.
SIMILARITY: Belongs to the annexin family. & Contains 4 annexin repeats.

Components

Component / Part No. / Quantity / Storage ApopNexin FITC / APT750a / 0.5 mL / 2-8°C Binding Buffer, 4X / APT750b / 50 mL / 2-8°C Propidium Iodide (PI), 20 μg/mL / APT750c / 1.6 mL / 2-8°C

Storage and Stability

ApopNexin FITC Apoptosis Detection Kit should be stored at 2-8°C and protected from light. Precautions · For research use only. Not for use in diagnostic procedures. · Part no. APT750a contains 0.02% sodium azide. · Part no. APT750c contains Propidium Iodide (PI), a suspected carcinogen. Handle with care using gloves and eye protection; do not ingest or contact to skin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service