Skip to Content
MilliporeSigma
All Photos(1)

Documents

APT115

Sigma-Aldrich

Apo-BrdU Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

species reactivity

all

Quality Level

technique(s)

flow cytometry: suitable

detection method

fluorometric

General description

The CHEMICON APO-BRDU Kit is a two color staining method for labeling DNA breaks and total cellular DNA to detect apoptotic cells by flow cytometry (Li et al. 1995). The kit contains the instructions and reagents required for measuring apoptosis in cells including; positive and negative control cells for assessing reagent performance; washing, reaction, and rinsing buffers for processing individual steps in the assay; terminal deoxynucleotidyl tranferase enzyme (TdT), bromodeoxyuridine triphosphate (Br-dUTP), and fluorescein labeled anti-BrdU antibody for labeling DNA breaks and Propidium Iodide/RNase A solution for counter staining the total DNA.

Components

The APO-BRDU Kit is shipped in one container and consists of two packages. One package is shipped at ambient temperature and should be stored at 2-8°C upon arrival. The other package is styrofoam containing frozen ice packs and the reagent contents should be stored at -20°C upon arrival. Note: Upon arrival store the reagents at the appropriate temperatures. Reagent bottles have color coded caps to aid in their identification. Sufficient reagents are provided to process 60 cell suspensions including 5 mL positive and 5 mL negative control cell suspensions of approximately 1 x 106 cells per mL in 70% (v/v) ethanol. The control cells are derived from a human lymphoma cell line and have been fixed as described on page 6. Positive Control Cells, Brown cap Negative Control Cells, White cap Wash Buffer, Blue cap Reaction Buffer, Green cap TdT Enzyme, Yellow cap Br-dUTP, Violet cap Rinsing Buffer, Red cap Fluorescein~PRB-1 mAb, Orange cap PI/RNase Staining Buffer, Amber bottle

Storage and Stability

Precautions and Warnings 1. The components of this kit are for Research Use only and are not intended for diagnostic procedures. 2. Positive and negative control cells contain 70% (v/v) ethanol as a preservative; wash and reaction buffer contain sodium cacodylate (dimethylarsinic) as a buffer; rinsing and PI/RNase staining buffer contain 0.05% (w/v) sodium azide as a preservative. These materials are harmful if swallowed; avoid skin contact, wash immediately with water. See Material Safety Data Sheets. 3. TdT Enzyme will not freeze at -20oC, because it is in 50% (v/v) glycerol solution. Upon warming the TdT enzyme solution, centrifuge the tube for 30 seconds to force all the liquid to the bottom of the tube.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service