Skip to Content
MilliporeSigma
All Photos(2)

Documents

AP30034P0

Millipore

Cellulose Fiber Membrane Filter

Millipore, filter diam. 34 mm, hydrophilic

Synonym(s):

Thick Absorbent Pad, 34 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24

material

cellulose fiber
plain filter
white filter

description

This is a 34 mm Hydrophilic Cellulose Fiber depth filter. Cellulose support pads are used to reinforce filters in monitors for contamination analysis. When saturated with growth medium, they can also be used for culturing microorganisms on membranes.

feature

hydrophilic

manufacturer/tradename

Millipore

filter diam.

34 mm

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Cellulose support pads are used to reinforce filters in Millipore′s monitors for contamination analysis. When saturated with growth medium, they can also be used for culturing microorganisms on membranes.

To increase flow rate and throughput, our woven mesh spacers can be placed between filters in a serial filtration to prevent the downstream screen filter from "blinding" the upstream filter pores.

Features & Benefits
•Avoid membrane damage during high pressure or fast flow conditions
•Spacers enable you to combine multiple stages of filtration into a single process step, saving time

Applications:
Air Monitoring, Environmental Monitoring, Aerosol Contamination Monitoring
Filter Code: AP30
Filter Type: Depth filter

Application

  • Air monitoring
  • Environmental monitoring
  • Aerosol contamination monitoring
  • Protecting membrane filters during high pressure or fast flow conditions


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service