Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AP194F

Sigma-Aldrich

Donkey Anti-Chicken IgY Antibody, FITC conjugate, Species Adsorbed

Chemicon®, from donkey

Synonym(s):

Chicken IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

donkey

Quality Level

conjugate

FITC conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

chicken

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Donkey anti-Chicken IgY Antibody, FITC conjugate, Species Adsorbed detects level of Donkey Chicken IgY & has been published & validated for use in IF.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suh Youn Ko et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 67(5), 335-349 (2018-12-24)
Netrin-1 is a potent axonal and neuronal guidance cue in the developing nervous system. Netrin-1 functions are mediated by its receptors, such as deleted in colorectal cancer (DCC) present on axons and neurons. Localization of DCC and Netrin-1 on various
Zahidul Khan et al.
International journal of molecular medicine, 28(6), 977-983 (2011-09-07)
Cell replacement and gene therapy using neural stem cells (NSCs) have been widely touted as a promising treatment for CNS diseases including brain tumors.
Arianna R S Lark et al.
Cellular and molecular neurobiology, 43(3), 1105-1127 (2022-06-14)
The striatum is especially vulnerable to HIV-1 infection, with medium spiny neurons (MSNs) exhibiting marked synaptodendritic damage that can be exacerbated by opioid use disorder. Despite known structural defects in MSNs co-exposed to HIV-1 Tat and opioids, the pathophysiological sequelae
Monocarboxylate transporter 1 (MCT1) plays a direct role in short-chain fatty acids absorption in caprine rumen.
Kirat, D; Masuoka, J; Hayashi, H; Iwano, H; Yokota, H; Taniyama, H; Kato, S
The Journal of Physiology null
Dong Kun Lee et al.
Nature communications, 6, 6618-6618 (2015-03-27)
Regulation of GABAergic inhibitory inputs and alterations in POMC neuron activity by nutrients and adiposity signals regulate energy and glucose homeostasis. Thus, understanding how POMC neurons integrate these two signal molecules at the synaptic level is important. Here we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service