Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AP180A

Sigma-Aldrich

Donkey Anti-Goat IgG Antibody, Alkaline Phosphatase conjugate, Species Adsorbed

0.6 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
$382.00

$382.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
500 μL
$382.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$382.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

donkey

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

goat

manufacturer/tradename

Chemicon®

concentration

0.6 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Related Categories

Specificity

Goat IgG (H+L) (absorbed for minimal cross-reactivity to Chicken, Guinea Pig, Syrian Hamster, Horse, Human, Mouse, Rabbit, and Rat Serum Proteins)

Application

ELISA and Western Blotting at 1:5000 - 1:50,000 dilution.Optimal working dilutions must be determined by the end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
This Donkey anti-Goat IgG Antibody, Alkaline Phosphatase conjugate, Species Adsorbed is validated for use in ELISA, WB for the detection of Donkey Goat IgG.

Physical form

Lyophilized. Buffer = 0.01 M Sodium Phosphate, 0.25 M NaCl, pH 8.0.

RECONSTITUTION:

Reconstitute to 1.0 mg/mL with 0.5 mL sterile distilled water.

Storage and Stability

Maintain lyophilized product at 2-8°C for up to 12 months. After reconstitution the product is stable for several weeks at 2-8°C as an undiluted liquid. After further dilution do not use for more than 1 day. For extended storage after reconstitution, add an equal volume of glycerol to make a final concentration of 50% glycerol followed by storage at -20°C in undiluted aliquots for up to 12 months. Please note the concentration of protein (and buffer salts) will decrease to one-half of the original after the addition of glycerol. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service