Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AP126P

Sigma-Aldrich

Chicken Anti-Mouse IgG Antibody, HRP conjugate

Chemicon®, from chicken

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.47

biological source

chicken

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Specificity

Specific for mouse IgG. One precipitin arc in immunodiffusion against mouse serum, dilution 1:10, or mouse IgG. Only one precipitin arc in immunoelectrophoresis against mouse serum or mouse IgG.

Immunogen

Mouse IgG

Application

Chicken anti-Mouse IgG Antibody, HRP conjugate detects level of Chicken Mouse IgG & has been published & validated for use in ELISA & WB.
ELISA: 1:70,000 dilution gave an O.D. of 1.0 in a 15 minute reaction using TMB as the substrate.

Optimal working dilutions must be determined by the end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Physical form

Affinity purified immunoglobulin conjugated with horseradish peroxidase. Liquid in 50 mM HEPES, pH 7.0, 0.1M NaCl, 0.2% BSA and 0.1% Kathon CG.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to six months after date of receipt. For long term storage dilute the antibody with sterile glycerol (ACS grade or better) and maintain at

-20°C for up to 12 months after date of receipt. Do not freeze the antibody unless it needs to be stored for an extended period of time. It is recommended that the antibody be stored in the dark.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuyan Wang et al.
Scientific reports, 10(1), 16725-16725 (2020-10-09)
Lubricin is an important boundary lubricant and chondroprotective glycoprotein in synovial fluid. Both increased and decreased synovial fluid lubricin concentrations have been reported in experimental post-traumatic osteoarthritis (PTOA) animal models and in naturally occurring joint injuries in humans and animals, with
Carolyn R Shurer et al.
Biotechnology and bioengineering, 116(6), 1292-1303 (2019-01-27)
Widespread therapeutic and commercial interest in recombinant mucin technology has emerged due to the unique ability of mucin glycoproteins to hydrate, protect, and lubricate biological surfaces. However, recombinant production of the large, highly repetitive domains that are characteristic of mucins
C Giaroni et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 25(2), e114-e126 (2013-01-03)
Intestinal ischemia and reperfusion (I/R) injury leads to abnormalities in motility, namely delay of transit, caused by damage to myenteric neurons. Alterations of the nitrergic transmission may occur in these conditions. This study investigated whether an in vitro I/R injury

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service