Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AGX812

Sigma-Aldrich

SARS-CoV-2 Nucleoprotein (aa 1-419) Recombinant Antigen, 613

suitable for immunodiffusion

Synonym(s):

2019-nCoV, COVID-19

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51201600
NACRES:
NA.26

recombinant

expressed in E. coli

Quality Level

form

liquid

mol wt

≤72 kDa

storage condition

avoid repeated freeze/thaw cycles

technique(s)

immunodiffusion: suitable
lateral flow assay: suitable

shipped in

dry ice

General description

SARS-CoV-2 Nucleoprotein (aa 1-419) Recombinant Antigen, expressed in E. coli, Cat. No. AGX812 is for use in Lateral Flow Assay.

Target description

Coronavirus disease 2019 (COVID-19) is an infectious disease caused by the virus strain severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus closely related to the SARS virus. A SARS-CoV-2 nucleoprotein antigen can be used to support the detection of SARS-CoV-2 (COVID-19, 2019-nCoV).

Physical form

Purified protein in 10mM Phosphate Buffered Saline without preservatives, pH 7.4.

Reconstitution

Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Stable for 1 year at -10°C to -25°C from date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service