Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AG918

Sigma-Aldrich

β Amyloid 1-40, aβ, F4W, TFA, recombinant human

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$685.00

$685.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$685.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

$685.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Quality Level

Assay

>97%

form

powder

manufacturer/tradename

Chemicon®

UniProt accession no.

shipped in

dry ice

Gene Information

human ... APP(351)

General description

Beta Amyloid (1-40), mutant, F4W, TFA

MOL. WT.: 4367.37 Da by Mass Spec Analysis (theoretical 4368.9)

SEQUENCE: Asp-Ala-Glu-Trp-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val

PEPTIDE CONTENT: 1 mg per vial; salt content is <10%

COUNTER ION: TFA

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service