Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AG382

Sigma-Aldrich

Uncoupling Protein 1, control peptide for AB3038

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.47

concentration

1 mg/mL

Quality Level

technique(s)

inhibition assay: suitable

UniProt accession no.

Gene Information

rat ... UCP1(24860)

General description

FUNCTION: SwissProt: P25874 # UCP are mitochondrial transporter proteins that create proton leaks across the inner mitochondrial membrane, thus uncoupling oxidative phosphorylation from ATP synthesis. As a result, energy is dissipated in the form of heat.
SIZE: 307 amino acids; 33005 Da
SUBUNIT: Acts as a dimer forming a proton channel.
SUBCELLULAR LOCATION: Mitochondrion inner membrane; Multi-pass membrane protein.
TISSUE SPECIFICITY: Brown adipose tissue.
SIMILARITY: SwissProt: P25874 ## Belongs to the mitochondrial carrier family. & Contains 3 Solcar repeats.

Application

Antibody blocking (AB3038)

Optimal working concentration must be determined by the end user.

Physical form

Liquid in PBS, pH 7.5 without a preservative

Storage and Stability

Store at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service