Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AG340

Sigma-Aldrich

Epithelial Sodium Channel γ, control peptide for AB3534P

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$248.00

$248.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$248.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$248.00


Please contact Customer Service for Availability

Request a Bulk Order

concentration

1 mg/mL

Quality Level

technique(s)

cell based assay: suitable

UniProt accession no.

Gene Information

General description

FUNCTION: SwissProt: P51170 # Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.
SIZE: 649 amino acids; 74270 Da
SUBUNIT: Heterotetramer of two alpha, one beta and one gamma subunit. A delta subunit can replace the alpha subunit. Interacts with the WW domains of NEDD4, NEDD4L, WWP1 and WWP2.
SUBCELLULAR LOCATION: Apical cell membrane; Multi-pass membrane protein. Note=Apical membrane of epithelial cells.
PTM: Phosphorylated on serine and threonine residues (By similarity). & Ubiquitinated; this targets individual subunits for endocytosis and proteasome-mediated degradation (By similarity).
DISEASE: SwissProt: P51170 # Defects in SCNN1G are a cause of Liddle syndrome [MIM:177200]. It is an autosomal dominant disorder characterized by pseudoaldosteronism and hypertension associated with hypokalemic alkalosis. The disease is caused by constitutive activation of the renal epithelial sodium channel.
SIMILARITY: SwissProt: P51170 ## Belongs to the amiloride-sensitive sodium channel family.

Application

Antibody blocking (AB3534P)

ELISA

Optimal working dilution must be determined by the end user.

Physical form

Liquid in PBS pH 7.4, containing 0.1% sodium azide.

Storage and Stability

Store at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service