Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AG290

Sigma-Aldrich

Opioid Receptor, µ, control peptide for AB1562

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

concentration

1 mg/mL

technique(s)

cell based assay: suitable

UniProt accession no.

Gene Information

mouse ... OPRM1(18390)

General description

FUNCTION: SwissProt: P35372 # Inhibits neurotransmitter release by reducing calcium ion currents and increasing potassium ion conductance. Receptor for beta-endorphin.
SIZE: 400 amino acids; 44779 Da
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
SIMILARITY: SwissProt: P35372 ## Belongs to the G-protein coupled receptor 1 family.

Application

Antibody blocking (AB1562).Care should be taken when diluting the peptide into buffer. It is suggested that you dilute by titrating buffer into a small amount of the peptide solution to avoid precipitation.

Optimal working concentration must be determined by the end user.

Physical form

Liquid in DMSO.

Storage and Stability

Store at room temperature in undiluted aliquots for up to 3 months.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service