Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AG231

Sigma-Aldrich

Neurofilament 70 kDa, porcine

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$403.00

$403.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
$403.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.41

$403.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

Porcine submaxillary glands

Quality Level

Assay

90% (NF-L and a small amount of NF-M)

form

liquid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

Porcine ... NEFL(100521224)

General description

Purified porcine axonal neurofilament 70 (low molecular weight, NF-L).

Application

Western blotting

Protein standard

ELISA

Radioimmunoassay

Optimal working dilution must be determined by the end user.

Physical form

Liquid in 6M Urea, 10 mM Phosphate, pH 7.5.

Storage and Stability

Maintain at -20°C to -70°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service